Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRB3 Monoclonal Antibody (N87/25)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA527698

Catalog No. PIMA527698


Only null left
Add to Cart

Description

Description

1 μg/mL of MA5-27698 was sufficient for detection of Beta 3 GABA receptor in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta1.

Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GABRB3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P28472, P63079, P63080
GABRB3
Fusion protein amino acids 370-433 of mouse GABA-A-R-Beta 3.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Microarray
N87/25
Unconjugated
GABRB3
A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; ECA5; GABA(A) receptor subunit beta-3; GABAA receptor beta-3 subunit; GABA-alpha receptor beta-2 subunit; GABR B3; Gabrb3; Gabrb-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA) A receptor, subunit beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit beta-3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3
Mouse
Protein G
RUO
14402, 24922, 2562
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.