Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115261
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115261 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115261 Supplier Thermo Scientific Supplier No. PA5115261
Add to Cart
Add to Cart

GABRB3 Polyclonal Antibody

GABRB3 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRB3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene GABRB3
Gene Accession No. P28472, P63079, P63080
Gene Alias A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; ECA5; GABA(A) receptor subunit beta-3; GABAA receptor beta-3 subunit; GABA-alpha receptor beta-2 subunit; GABR B3; Gabrb3; Gabrb-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA) A receptor, subunit beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit beta-3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3
Gene Symbols GABRB3
Host Species Rabbit
Immunogen A synthesized peptide derived from human GABRB3(Accession P28472), corresponding to amino acid residues N366-L416.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14402, 24922, 2562
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.