Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597766
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597766 Supplier Invitrogen™ Supplier No. PA597766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GABRB3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Gamma-amino butyric acid (GAMA) is a strong inhibitor of neuron transmission and is found throughout the central nervous system. GABA is formed by the decarboxylation of glutamate, resulting in the formation of one of two distinct types; GABA-A or GABA-B.The binding of GABA to GABA-A receptors results in the increase transportation of chlorine ions into the neuron. This influx results in signal transmission repression. Studies have shown that if cell to cell signaling goes unchecked due to a lack of GABA, a system overload can resulting in blackouts, rapid body contractions, seizures and epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRB3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GABRB3
Gene Accession No. P28472
Gene Alias A230092K12Rik; AW049585; beta3; cleft palate 1; Cp1; ECA5; GABA(A) receptor subunit beta-3; GABAA receptor beta-3 subunit; GABA-alpha receptor beta-2 subunit; GABR B3; Gabrb3; Gabrb-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, beta 3; gamma-aminobutyric acid (GABA) A receptor, subunit beta 3; gamma-aminobutyric acid (GABA-A) receptor, subunit beta 3; gamma-aminobutyric acid A receptor beta 3; Gamma-aminobutyric acid receptor beta 3; gamma-aminobutyric acid receptor subunit beta-3; gamma-aminobutyric acid receptor, subunit beta 3; gamma-aminobutyric acid type A receptor beta3 subunit; gamma-aminobutyric acid type A receptor subunit beta3; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit beta-3; MGC9051; testis gamma-aminobutyric acid receptor subunit beta 3
Gene Symbols GABRB3
Host Species Rabbit
Immunogen Recombinant Human Gamma-aminobutyric acid receptor subunit beta-3 protein (328-450AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2562
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.