Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRD Monoclonal Antibody (N151/3), PE
GREENER_CHOICE

Catalog No. PIMA545484
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545484 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545484 Supplier Invitrogen™ Supplier No. MA545484
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

2 μg/mL of MA5-45484 was sufficient for detection of Delta1 GABA-A receptor in 10 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 55kDa.

GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRD
Applications Immunohistochemistry, Western Blot, Immunocytochemistry, Microarray
Classification Monoclonal
Clone N151/3
Concentration 1 mg/mL
Conjugate PE
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene GABRD
Gene Accession No. O14764, P18506, P22933
Gene Alias AI853201; EIG10; EJM7; GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABAA receptor delta subunit; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA-RD; GABAARdelta; Gabrd; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, subunit delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid receptor subunit delta; gamma-aminobutyric acid type A receptor delta subunit; GEFSP5; MGC105467; MGC45284
Gene Symbols GABRD
Host Species Mouse
Immunogen Synthetic peptide amino acids 15-34 (N-terminus) of rat GABA-A-R-Delta.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14403, 2563, 29689
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.