Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101598
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101598 Supplier Invitrogen™ Supplier No. PA5101598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GABRD.

GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GABRD
Gene Accession No. O14764, P18506, P22933
Gene Alias AI853201; EIG10; EJM7; GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABAA receptor delta subunit; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA-RD; GABAARdelta; Gabrd; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, subunit delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid receptor subunit delta; gamma-aminobutyric acid type A receptor delta subunit; GEFSP5; MGC105467; MGC45284
Gene Symbols GABRD
Host Species Rabbit
Immunogen A synthesized peptide derived from human GABRD(Accession O14764), corresponding to amino acid residues D149-S199.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14403, 2563, 29689
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.