Learn More
Invitrogen™ GABRD Polyclonal Antibody

Description
GABRD Polyclonal Antibody for Flow, IHC (P), Western Blot
Specifications
Specifications
| Antigen | GABRD |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GABRD |
| Gene Accession No. | O14764 |
| Gene Alias | AI853201; EIG10; EJM7; GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABAA receptor delta subunit; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA-RD; GABAARdelta; Gabrd; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, subunit delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid receptor subunit delta; gamma-aminobutyric acid type A receptor delta subunit; GEFSP5; MGC105467; MGC45284 |
| Gene Symbols | GABRD |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.