Learn More
Invitrogen™ GABRE Monoclonal Antibody (S413-67)
Description
GABRE Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Specifications
Specifications
| Antigen | GABRE |
| Applications | Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | S413-67 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 |
| Gene | Gabre |
| Gene Accession No. | P78334, Q9ES14 |
| Gene Alias | GABA(A) mu Receptor; GABA(A) receptor; GABA(A) receptor subunit epsilon; GABA(A) receptor, epsilon; GABRE; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, epsilon; gamma-aminobutyric acid (GABA) A receptor, subunit epsilon; gamma-aminobutyric acid (GABA-A) receptor, subunit epsilon; gamma-aminobutyric acid A receptor, epsilon; gamma-aminobutyric acid receptor subunit epsilon; gamma-aminobutyric acid type A receptor epsilon subunit; gamma-aminobutyric acid type A receptor subunit epsilon |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.