Learn More
Invitrogen™ GABRG2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine and rat based on sequence homology.
Specifications
Specifications
| Antigen | GABRG2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Gabrg2 |
| Gene Accession No. | P18507, P22723 |
| Gene Alias | CAE2; ECA2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; GABAA-R; GABRA2; Gabrg2; Gabrg-2; gamma2; gamma-aminobutyrate receptor type A gamma-2 subuni; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor gamma 2; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit; gamma-aminobutyric acid type A receptor subunit gamma2; GEFSP3; LOC553402 protein; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit gamma-2; si:ch211-145n14.1 |
| Gene Symbols | Gabrg2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.