Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541331
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541331 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541331 Supplier Invitrogen™ Supplier No. PA541331
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VPEGDVTVIL NNLLEGYDNK LRPDIGVKPT LIHTDMYVNS IGPVNAINME Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRG2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gabrg2
Gene Accession No. P18507
Gene Alias CAE2; ECA2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; GABAA-R; GABRA2; Gabrg2; Gabrg-2; gamma2; gamma-aminobutyrate receptor type A gamma-2 subuni; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor gamma 2; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit; gamma-aminobutyric acid type A receptor subunit gamma2; GEFSP3; LOC553402 protein; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit gamma-2; si:ch211-145n14.1
Gene Symbols Gabrg2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GABRG2 (aa 61-110).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2566
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.