Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABRG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571457
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a gamma-aminobutyric acid (GABA) receptor. GABA is the major inhibitory neurotransmitter in the mammlian brain, where it acts at GABA-A receptors, which are ligand-gated chloride channels. GABA-A receptors are pentameric, consisting of proteins from several subunit classes: alpha, beta, gamma, delta and rho. Mutations in this gene have been associated with epilepsy and febrile seizures. Multiple transcript variants encoding different isoforms have been identified for this gene.
Specifications
GABRG2 | |
Polyclonal | |
Unconjugated | |
Gabrg2 | |
CAE2; ECA2; GABA(A) receptor subunit gamma-2; GABA(A) receptor, gamma 2; GABAA-R; GABRA2; Gabrg2; Gabrg-2; gamma2; gamma-aminobutyrate receptor type A gamma-2 subuni; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, gamma 2; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 2; gamma-aminobutyric acid A receptor gamma 2; gamma-aminobutyric acid A receptor, gamma 2; gamma-aminobutyric acid receptor subunit gamma-2; gamma-aminobutyric acid type A receptor gamma2 subunit; gamma-aminobutyric acid type A receptor subunit gamma2; GEFSP3; LOC553402 protein; LOW QUALITY PROTEIN: gamma-aminobutyric acid receptor subunit gamma-2; si:ch211-145n14.1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2566 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P18507 | |
Gabrg2 | |
synthetic peptide directed towards the N terminal region of human GABRG2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction