Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589095
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589095 Supplier Invitrogen™ Supplier No. PA589095
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NYLFYSRGPR RQPRRHRRPR RVIARYRYQQ VVVGNVQDGL INVEDGVSSL PITPAQAPLA SPESLGSLTS TSEQAQLATS ESLSPLTSLS GQAPLATGES LSDLPSTSEQ ARHSYGVRFN GFQADDSIIP TEIRNRVEAH GHGVTHDHED SNESLSSDER HGHGPSGKPM LHHGEKGVQE AGWDLDDNND KSDCLAIKEQ FKCDTNSTWG LNDDELMAHG QEKDSSSESE DSCPPSPGCS FTEGFSFDLF N; Positive Samples: Jurkat; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, postsynaptic cell membrane, synapse.

The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. GABRQ gene is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases:early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).The gamma-aminobutyric acid (GABA) A receptor is a multisubunit chloride channel that mediates the fastest inhibitory synaptic transmission in the central nervous system. This gene encodes GABA A receptor, theta subunit. It is mapped to chromosome Xq28 in a cluster including the genes encoding the alpha 3 and epsilon subunits of the same receptor. This gene location is also the candidate region of 2 different neurologic diseases: early-onset parkinsonism (Waisman syndrome) and X-linked mental retardation (MRX3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRQ
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GABRQ
Gene Accession No. Q9UN88
Gene Alias GABA(A) receptor subunit theta; GABA(A) Theta; GABRQ; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid (GABA) A receptor, subunit theta; gamma-aminobutyric acid (GABA) A receptor, theta; gamma-aminobutyric acid (GABA) receptor, theta; gamma-aminobutyric acid (GABA-A) receptor, subunit theta; gamma-aminobutyric acid A receptor, theta; gamma-aminobutyric acid receptor subunit theta; gamma-aminobutyric acid type A receptor subunit theta; gamma-aminobutyric acid type A receptor theta subunit; THETA
Gene Symbols GABRQ
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-600 of human GABRQ (NP_0610283).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55879
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.