Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103849 Supplier Invitrogen™ Supplier No. PA5103849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GABRR1.

GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels; however, they are structurally and functionally distinct. GABAC receptors (GABAC Rr) mediate rapid inhibitory neurotransmission in retina. Three human genes, r1 (GABRR1), r2 (GABRR2) and r3 (GABRR3), encode the three polypeptides that comprise this receptor. GABRR1 and GABRR2 are located close together, in a region of chromosome 6q that contains loci for inherited disorders of the eye, but GABRR3 maps to chromosome 3q11-q13.3. The r polypeptide genes, which are thought to share a common ancestor with GABA(A) receptor subunit genes, diverged at an early stage in the evolution of this gene family. The expression of GABAC Rr subunits is not restricted to the retina, but significant expression can also be detected in many other brain regions, especially in those belonging to the visual pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRR1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GABRR1
Gene Accession No. P24046
Gene Alias bA135P14.1 (gamma-aminobutyric acid (GABA) receptor, rho 1); GABA(A) receptor subunit rho-1; GABA(A) receptor, rho 1; GABA(C) receptor; GABA(C) rho1; GABA-C; Gabrr1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 1; gamma-aminobutyric acid (GABA) A receptor, rho-1; gamma-aminobutyric acid (GABA) C receptor, subunit rho 1; gamma-aminobutyric acid (GABA) receptor, rho 1; gamma-aminobutyric acid (GABA-A) receptor subunit rho 1; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 1; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 1; gamma-aminobutyric acid A receptor, rho 1; gamma-aminobutyric acid receptor subunit rho-1; gamma-aminobutyric acid type A receptor rho1 subunit
Gene Symbols GABRR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GABRR1(Accession P24046), corresponding to amino acid residues S213-S263.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2569
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.