Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GABRR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112424
Description
GABRR1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels; however, they are structurally and functionally distinct. GABAC receptors (GABAC Rr) mediate rapid inhibitory neurotransmission in retina. Three human genes, r1 (GABRR1), r2 (GABRR2) and r3 (GABRR3), encode the three polypeptides that comprise this receptor. GABRR1 and GABRR2 are located close together, in a region of chromosome 6q that contains loci for inherited disorders of the eye, but GABRR3 maps to chromosome 3q11-q13.3. The r polypeptide genes, which are thought to share a common ancestor with GABA(A) receptor subunit genes, diverged at an early stage in the evolution of this gene family. The expression of GABAC Rr subunits is not restricted to the retina, but significant expression can also be detected in many other brain regions, especially in those belonging to the visual pathways.
Specifications
| GABRR1 | |
| Polyclonal | |
| Unconjugated | |
| GABRR1 | |
| bA135P14.1 (gamma-aminobutyric acid (GABA) receptor, rho 1); GABA(A) receptor subunit rho-1; GABA(A) receptor, rho 1; GABA(C) receptor; GABA(C) rho1; GABA-C; Gabrr1; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 1; gamma-aminobutyric acid (GABA) A receptor, rho-1; gamma-aminobutyric acid (GABA) C receptor, subunit rho 1; gamma-aminobutyric acid (GABA) receptor, rho 1; gamma-aminobutyric acid (GABA-A) receptor subunit rho 1; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 1; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 1; gamma-aminobutyric acid A receptor, rho 1; gamma-aminobutyric acid receptor subunit rho-1; gamma-aminobutyric acid type A receptor rho1 subunit | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14408, 2569 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P24046, P56475 | |
| GABRR1 | |
| Recombinant Human Gamma-aminobutyric acid receptor subunit rho-1 protein (365-465AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction