Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GABRR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529472
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529472 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529472 Supplier Invitrogen™ Supplier No. PA529472
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: SK-N-SH, Neuro2A. Predicted reactivity: Mouse (97%), Rat (97%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA receptors, which are ligand-gated chloride channels. The protein encoded by this gene is a member of the rho subunit family and is a component of the GABA receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene Gabrr2
Gene Accession No. P28476, P56476
Gene Alias GABA(A) receptor; GABA(A) receptor subunit rho-2; GABA(C) receptor; GABA-C receptor, rho-2 subunit; Gabrr2; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 2; gamma-aminobutyric acid (GABA) C receptor, subunit rho 2; gamma-aminobutyric acid (GABA) receptor, rho 2; gamma-aminobutyric acid (GABA-A) receptor subunit rho 2; gamma-aminobutyric acid (GABA-A) receptor, subunit rho 2; gamma-aminobutyric acid (GABA-C) receptor, subunit rho 2; gamma-aminobutyric acid A receptor, rho 2; gamma-aminobutyric acid receptor rho2 subunit; gamma-aminobutyric acid receptor subunit rho-2; gamma-aminobutyric acid type A receptor rho2 subunit; gamma-aminobutyric acid type A receptor subunit rho2
Gene Symbols Gabrr2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GABRR2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14409, 2570
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.