Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABRR3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548371
Description
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
Specifications
GABRR3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
A8MPY1 | |
GABRR3 | |
KLH conjugated synthetic peptide between 35-63 amino acids from the N-terminal region of human GBRR3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GABRR3 | |
GABA receptor rho-3 subunit; GABA(A) receptor subunit rho-3; GABA(C) receptor; GABRR3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, rho 3; gamma-aminobutyric acid (GABA) receptor, rho 3; gamma-aminobutyric acid receptor subunit rho-3; gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
200959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction