Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD 65/67, Rabbit anti-Human, Mouse, Rat, Polyclonal, Millipore Sigma™

Catalog No. ABN90425UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN90425UG 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN90425UG Supplier MilliporeSigma Supplier No. ABN90425UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GAD 65/67 Antibody, Cat. No. ABN904, is a highly specific rabbit polyclonal antibody that targets Human, Mouse, and Rat GAD2 and GAD1 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.

Glutamate decarboxylase 2/Glutamate decarboxylase 1 (UniProt: Q05329/Q99259; also known as 65 kDa/67kDa glutamic acid decarboxylase, GAD-65/67, Glutamate decarboxylase 65/67 kDa isoform) is encoded by GAD2/GAD1 (also known as GAD65/GAD67) gene (Gene ID: 2572/2571) in human. Glutamate decarboxylase isoforms are expressed in the brain where it is involved in gamma-aminobutyric acid (GABA) synthesis. Glutamate decarboxylase 2 is also expressed in pancreatic tissue. Both GAD65 and GAD67 synthesize GABA at different locations in the cell and at different developmental stages. Usually, GAD1 is distributed evenly throughout the cell while isoform GAD2 is mainly localized to nerve terminals. GAD1 synthesizes GABA for neuronal activity that is related to as synaptogenesis and protection from neural injury. However, GAD2 isoform synthesizes GABA for neurotransmission activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAD 65/67
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected GAD 65/67 in human cerebellum, mouse cerebral cortex, and rat cerebral cortex tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Glutamate decarboxylase 2/Glutamate decarboxylase 1;65 kDa/67kDa glutamic acid decarboxylase;GAD-65/67;Glutamate decarboxylase 65/67 kDa isoform
Gene Symbols GAD2/GAD1;GAD65/GAD67
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human glutamate decarboxylase 2 (GAD2).
Molecular Weight of Antigen ∽60/62kDa observed; 65.41/66.90 kDa calculated, respectively for GAD2 and GAD1. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects GAD2 and GAD1 in human, Mouse, and Rat. It targets an epitope within 14 amino acids from the C-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.