Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Antibody (CL2914), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$382.00

Specifications

Antigen GAD1/GAD67
Clone CL2914
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB396377
SDP
View Documents
Novus Biologicals
NBP24663825UL
25 μL
Each of 1 for $382.00
Only null left
Add to Cart
 
Description

Description

GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

GAD1/GAD67
Western Blot 1 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Monoclonal
Purified
RUO
Human, Mouse, Rat
Q99259
2571
IgG1
Protein A purified
CL2914
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Diabetes Research
PBS (pH 7.2), 40% Glycerol
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence QSSKNLLSCENSDRDARFRRTETDFSNLFARDLLPAKNGEEQTVQFLLEVVDILLNYVRKTFDRS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.