Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67 Antibody (GAD1/2563), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279937MFV450
Description
GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human samples. It is validated for Flow CytometrySpecifications
GAD1/GAD67 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
GAD1/2563 | |
Flow Cytometry | |
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) (Uniprot: Q99259) | |
0.1 mL | |
Diabetes Research | |
2571 | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction