Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67 Antibody (OTI3G9), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270791FR
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GAD1/GAD67 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
GAD1 | |
Full length recombinant protein of human GAD1/GAD67 (NP_000808) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3G9 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
Mouse | |
Affinity Purified | |
Diabetes Research | |
2571.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction