Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67 Antibody (OTI3G9), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP148272
Description
GAD1/GAD67 Monoclonal specifically detects GAD1/GAD67 in Human, Mouse, Rat, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
GAD1/GAD67 | |
Monoclonal | |
0.46 mg/mL | |
Western Blot 1:1000-2000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 4 ug/mL, Immunohistochemistry-Paraffin 1:50, Peptide ELISA | |
GAD1 | |
Full length human recombinant protein of human GAD1 (NP_000808) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens glutamate decarboxylase 1 (brain, 67kDa) (GAD1), transcript variant GAD67. | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
OTI3G9 | |
Unconjugated | |
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
Mouse | |
66.9 kDa | |
0.1 mL | |
Diabetes Research | |
2571 | |
Human, Mouse, Rat, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction