Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67 Rabbit anti-Human, Mouse, Rat, Clone: 4I8S8, Novus Biologicals™

Rabbit Monoclonal Antibody
$196.25 - $452.15
Specifications
| Antigen | GAD1/GAD67 |
|---|---|
| Clone | 4I8S8 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| GAD1/GAD67 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Diabetes Research | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2571 | |
| IgG | |
| Affinity purified |
| 4I8S8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human GAD1/GAD67 (Q99259). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title