Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1 Mouse anti-Human, Unconjugated, Clone: GAD1/2391, Abnova™

Catalog No. 89337692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-692 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-692 Supplier Abnova Corporation Supplier No. MAB21199
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GAD1 Monoclonal antibody specifically detects GAD1 in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen GAD1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GAD1/2391
Conjugate Unconjugated
Description glutamate decarboxylase 1 (brain, 67kDa)
Gene GAD1
Gene Alias FLJ45882, GAD, SCP
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 72-135 of human GAD1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2571
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.