Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD2 Mouse anti-Human, Unconjugated, Clone: GAD2/2362, Abnova™

Catalog No. 89337496 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-496 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-496 Supplier Abnova Corporation Supplier No. MAB21003
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GAD2 Monoclonal antibody specifically detects GAD2 in Human samples. It is validated for ELISA, WB-Ce, IHC-P.

Specifications

Antigen GAD2
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GAD2/2362
Conjugate Unconjugated
Description glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa)
Gene GAD2
Gene Alias GAD65, MGC161605, MGC161607
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 6-99 of human GAD2
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2572
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.