Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GAD65/GAD67 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104543
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104543 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104543 Supplier Invitrogen™ Supplier No. PA5104543
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GAD1/2.

GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA ( gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predominant GAD-67 form and the minor GAD-25 form.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAD65/GAD67
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gad1
Gene Accession No. P18088, P48318, P48320, Q05329, Q05683, Q99259
Gene Alias 6330404F12Rik; 65 kDa glutamic acid decarboxylase; 67 kDa glutamic acid decarboxylase; CPSQ1; EC 4.1.1.15; EP10; FLJ45882; GAD; GAD 1; GAD 2; GAD 65; GAD 67; GAD(65); GAD1; Gad-1; Gad2; Gad-2; GAD25; GAD44; Gad65; GAD-65; GAD67; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); glutamate decarboxylase 1 variant GAD67NT; glutamate decarboxylase 2; Glutamate decarboxylase 2 (islet); glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa); glutamate decarboxylase 65 kDa isoform; Glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); Glutamate decarboxylase-2 (pancreas); glutamic acid decarboxylase; glutamic acid decarboxylase 1; glutamic acid decarboxylase 2; glutamic acid decarboxylase 65; SCP
Gene Symbols Gad1, GAD2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GAD1(Accession Q99259), corresponding to amino acid residues D561-L594.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14415, 14417, 24379, 24380, 2571, 2572
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.