Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD67 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA584379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584379 Supplier Thermo Scientific Supplier No. PA584379

Rabbit Polyclonal Antibody

Immunogen sequence: QSSKNLLSCE NSDRDARFRR TETDFSNLFA RDLLPAKNGE EQTVQFLLEV VDILLNYVRK TFDRS

Glutamic Acid Decarboxylase (GAD) catalyzes the conversion of L-glutamate to gamma-aminobutyric (GABA), the principal inhibitory neurotransmitter in the brain, and a putative paracrine signal molecule in pancreatic islets. GAD67 is a cytoplasmic protein and is encodes on chromosome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GAD67
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAD1
Gene Accession No. Q99259
Gene Alias GAD
Gene Symbols GAD1
Host Species Rabbit
Immunogen Recombit protein corresponding to Human GAD1
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2571
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.