Learn More
Invitrogen™ GAD67 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585371
Description
Keep as concentrated solution. Predicted reactivity: Mouse (96%), Rat (96%), Dog (99%), Cat (99%), Pig (99%), Chicken (91%), Chimpanzee (100%), Bovine (98%). Positive Control: SK-N-SH, mouse brain, rat brain, human GAD1-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA (gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predominant GAD-67 form and the minor GAD-25 form.
Specifications
GAD67 | |
Polyclonal | |
Unconjugated | |
GAD1 | |
67 kDa glutamic acid decarboxylase; 67kDa glutamate decarboxylase 1; CPSQ1; EGK_04525; EP10; FLJ45882; GAD; GAD 1; GAD 67; GAD1; Gad-1; GAD25; GAD44; Gad67; GAD-67; Glutamate decarboxylase 1; Glutamate decarboxylase 1 (brain); glutamate decarboxylase 1 (brain, 67kDa); glutamate decarboxylase 1 isoform GAD67; glutamate decarboxylase 1 variant GAD67NT; glutamate decarboxylase 67 kDa isoform; GLUTAMATE DECARBOXYLASE, 67 KD ISOFORM (GAD-67) (67 KD GLUTAMIC ACID DECARBOXYLASE); glutamic acid decarboxylase; glutamic acid decarboxylase 1; glutamic acid decarboxylase isoform 67; OTTHUMP00000041055; SCP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14415, 24379, 2571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.17 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P18088, P48318, Q99259 | |
GAD1 | |
Recombinant protein encompassing a sequence within the center region of human GAD67. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.