Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD153/CHOP Antibody (17A10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | GADD153/CHOP |
---|---|
Clone | 17A10 |
Dilution | Flow Cytometry 1:50-1:200, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
GADD153/CHOP Monoclonal antibody specifically detects GADD153/CHOP in Human samples. It is validated for Flow Cytometry, ELISASpecifications
GADD153/CHOP | |
Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
C/EBP-homologous protein, C/EBP-homologous protein 10, CCAAT/enhancer-binding protein homologous protein, CHOP-10, CHOP10 CEBPZ, CHOPC/EBP zeta, DDIT-3, DNA damage-inducible transcript 3 protein, DNA-damage-inducible transcript 3, GADD153, Growth arrest and DNA damage-inducible protein GADD153 | |
A synthesized peptide derived from Human GADD153/CHOP [UniProt P35638] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
17A10 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis, Cell Cycle and Replication, DNA Repair, Hypoxia, Lipid and Metabolism, Neuroscience, Unfolded Protein Response | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1649 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title