Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD153/DDIT3 Polyclonal Antibody, PE, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS20669RPE

This item is not returnable.
View return policy
Description
GADD153/DDIT3 Polyclonal antibody specifically detects GADD153/DDIT3 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
GADD153/DDIT3 | |
Polyclonal | |
PE | |
DDIT3 | |
C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; C/EBP-homologous protein; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; CEBPZ; CHOP; CHOP10; CHOP-10; Ddit3; DDIT-3; ddit3.L; DNA damage inducible transcript 3; DNA damage inducible transcript 3 L homeolog; DNA damage-inducible transcript 3 protein; DNA-damage inducible transcript 3; DNA-damage-inducible transcript 3; GADD153; growth arrest and DNA damage-inducible; growth arrest and DNA damage-inducible protein GADD153; Growth arrest and DNA-damage-inducible protein GADD153; MGC4154; transcription factor GADD153; Xchop; XELAEV_18013440mg | |
KLH conjugated synthetic peptide derived from human GADD153/DDIT3 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
P35638, P35639, Q62857 | |
Rabbit | |
Protein A | |
RUO | |
13198, 1649, 29467 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction