Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD153 Polyclonal Antibody, Abbomax

Catalog No. 502204072
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4072 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4072 Supplier Abbomax Inc Supplier No. 602430
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: NIH3T3 cell lysate Cellular location: Nucleus.

GADD153 Polyclonal antibody specifically detects GADD153 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GADD153
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene DDIT3
Gene Accession No. P35638
Gene Alias C/EBP homologous protein; C/EBP homoologous protein 10; C/EBP zeta; C/EBP-homologous protein; C/EBP-homologous protein 10; CCAAT/enhancer-binding protein homologous protein; CEBPZ; CHOP; CHOP10; CHOP-10; Ddit3; DDIT-3; ddit3.L; DNA damage inducible transcript 3; DNA damage inducible transcript 3 L homeolog; DNA damage-inducible transcript 3 protein; DNA-damage inducible transcript 3; DNA-damage-inducible transcript 3; GADD153; growth arrest and DNA damage-inducible; growth arrest and DNA damage-inducible protein GADD153; Growth arrest and DNA-damage-inducible protein GADD153; MGC4154; transcription factor GADD153; Xchop; XELAEV_18013440mg
Host Species Rabbit
Immunogen A synthetic peptide of human ID3
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.