Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GADD34 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117768
This item is not returnable.
View return policy
Description
GADD34 Polyclonal Antibody for Western Blot, ICC/IF, IP
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.
Specifications
| GADD34 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R15A | |
| 9630030H21; Gadd34; growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; Myd116; myeloid differentiation primary response gene 116; myeloid differentiation primary response protein MyD116; Myeloid differentiation primary response protein MyD116 homolog; Peg3; PEG-3; PPP1R15A; PR15A; Progression elevated gene 3 protein; protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A; RGD1624209 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23645 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.38 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O75807 | |
| PPP1R15A | |
| E. coli-derived Human ADD3 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction