Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD34 Rabbit anti-Bovine, Human, Non-human Primate, Panda, Polyclonal, FabGennix
Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc GADD34101AP

Description
GADD34 Polyclonal antibody specifically detects GADD34 in Bovine, Human, Non-human Primate, Panda samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
GADD34 | |
Polyclonal | |
Unconjugated | |
PPP1R15A | |
9630030H21; Gadd34; growth arrest and DNA damage-inducible protein GADD34; growth arrest and DNA-damage-inducible 34; Myd116; myeloid differentiation primary response gene 116; myeloid differentiation primary response protein MyD116; Myeloid differentiation primary response protein MyD116 homolog; Peg3; PEG-3; PPP1R15A; PR15A; Progression elevated gene 3 protein; protein phosphatase 1 regulatory subunit 15A; protein phosphatase 1, regulatory (inhibitor) subunit 15A; protein phosphatase 1, regulatory subunit 15A; RGD1624209 | |
Synthetic peptide corresponding to internal sequence of amino acids 380-400 of Human GADD34, covalently modified and coupled to a carrier protein.Sequence:PATGVFLKSWVYQPGEDTEEE | |
200 μL | |
Primary | |
Bovine, Human, Non-human Primate, Panda | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
O75807, Q2KI51 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100471595, 23645, 456191, 514688 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction