Learn More
GADD34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GADD34 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-556-111
![]() ![]() |
Proteintech Group Inc
104491AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Molecular weight of GADD34 is 100 kDa confirmed in GADD34 KO mice, and Proteintech's GADD34 antibody 10449-1-AP primarily recognize the 100 kDa band.
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.Specifications
GADD34 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O75807, P17564, Q6IN02 | |
171071, 17872, 23645 | |
GADD34 Fusion Protein Ag0578 | |
Primary | |
-20°C | |
PPP1R15A |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GADD34, growth arrest and DNA damage-inducible protein 34, PPP1R15A | |
Ppp1r15a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.