Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45 alpha Antibody, Novus Biologicals™
SDP

Catalog No. NBP179791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP179791 100 μL
NBP1797920 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP179791 Supplier Novus Biologicals Supplier No. NBP179791
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GADD45 alpha Polyclonal specifically detects GADD45 alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen GADD45 alpha
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_001915
Gene Alias DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha
Gene Symbols GADD45A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal of human GADD45AThe immunogen for this antibody is GADD45A. Peptide sequence LRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPA.
Molecular Weight of Antigen 18 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1647
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Pig, Bovine, Equine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.