Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD45 alpha Antibody (OTI1C9), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270559V
Description
GADD45 alpha Monoclonal specifically detects GADD45 alpha in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GADD45 alpha | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GADD45A | |
Full length recombinant protein of human GADD45 alpha (NP_001915) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot | |
OTI1C9 | |
Western Blot | |
DDIT-1, DDIT1growth arrest and DNA damage-inducible protein GADD45 alpha, DNA damage-inducible transcript 1 protein, DNA damage-inducible transcript-1, GADD45DNA-damage-inducible transcript 1, growth arrest and DNA-damage-inducible 45 alpha, growth arrest and DNA-damage-inducible, alpha | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer | |
1647.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction