Learn More
GADD45A Mouse anti-Human, Clone: OTI2B7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF507380

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GADD45 alpha, beta and gamma are an evolutionarily conserved, homologous family of nuclear proteins that function as stress sensors for cellular physiological and environmental damage. GADD45 proteins are required for the activation of the G2/M checkpoint induced by UV radiation or alkylating agents. GADD45-induced G2/M checkpoint is regulated through inactivation of Cdc2-cyclin B1 kinase. GADD45 forms a complex with p21 and Cdc2, and may serve as core for interaction with other cell cycle regulators.Specifications
GADD45A | |
Monoclonal | |
Unconjugated | |
GADD45A | |
DDIT1, GADD45 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1647 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2B7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P24522 | |
GADD45A | |
Full length human recombit protein of human GADD45A produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.