Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GADD45A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565786 Supplier Invitrogen™ Supplier No. PA565786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTLEEFSAGEQ KTERMDKVGD ALEEVLSKAL SQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 88%.

GADD45 alpha, beta and gamma are an evolutionarily conserved, homologous family of nuclear proteins that function as stress sensors for cellular physiological and environmental damage. GADD45 proteins are required for the activation of the G2/M checkpoint induced by UV radiation or alkylating agents. GADD45-induced G2/M checkpoint is regulated through inactivation of Cdc2-cyclin B1 kinase. GADD45 forms a complex with p21 and Cdc2, and may serve as core for interaction with other cell cycle regulators.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GADD45A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GADD45A
Gene Accession No. P24522
Gene Alias AA545191; DDIT 1; DDIT1; DDIT-1; DNA damage-inducible transcript 1 protein; DNA damage-inducible transcript-1; DNA-damage inducible transcript 1; DNA-damage-inducible transcript 1; GADD45; GADD45A; Gadd45alpha; growth arrest and DNA damage inducible alpha; growth arrest and DNA damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible 45 alpha; growth arrest and DNA-damage-inducible protein GADD45 alpha; growth arrest and DNA-damage-inducible, alpha
Gene Symbols GADD45A
Host Species Rabbit
Immunogen Recombinant Human GADD45A. Recombinant protein control fragment (Product #RP-106462).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.