Learn More
Invitrogen™ GADD45B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143903
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MCF-7 whole cell. IHC: human placenta tissue, human spleen tissue, human renal cell carcinoma tissue, human tonsil tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Cellcycle progression issubject to arrest at G1 and G2 checkpointsin response to DNA damage, presumablyto allow time for DNA repair prior to entryinto S andMphase, respectively. The p53 tumorsuppressor isrequired for one such G1 checkpoint and functionsto upregulate expression of GADD 45 and p21. GADD 45 binds both Cdks and PCNA, a protein involved in DNA replication and repair. GADD 45 stimulates DNA excision repair in vitro and inhibits entry ofcellsinto S phase. Thus, it has been suggested that GADD 45 mayserve as a link between the p53-dependentcellcycle checkpoint and DNA repair. GADD 45-like proteins, GADD 45 beta and GADD 45 gamma, have been shown to be induced by environmentalstresses. GADD 45 beta and GADD 45 gamma are thought to induce p38/JNK activation via MEKK4 activation.
Specifications
GADD45B | |
Polyclonal | |
Unconjugated | |
GADD45B | |
AI323528; DKFZP566B133; Gadd45b; GADD45BETA; growth arrest and DNA damage inducible beta; growth arrest and DNA damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible 45 beta; growth arrest and DNA-damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible, beta; MYD118; myeloid differentiation primary response; myeloid differentiation primary response gene 118; myeloid differentiation primary response protein MyD118; Negative growth regulatory protein MyD118; Unknown (protein for MGC:138134) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4616 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
O75293 | |
GADD45B | |
A synthetic peptide corresponding to a sequence of human GADD45B (EELLVAAQRQDRLTVGVYE). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.