Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GADD45B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543160
Description
Peptide sequence: FCCDNDINIV RVSGMQRLAQ LLGEPAETQG TTEARDLHCL LVTNPHTDAW Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Cellcycle progression issubject to arrest at G1 and G2 checkpointsin response to DNA damage, presumablyto allow time for DNA repair prior to entryinto S andMphase, respectively. The p53 tumorsuppressor isrequired for one such G1 checkpoint and functionsto upregulate expression of GADD 45 and p21. GADD 45 binds both Cdks and PCNA, a protein involved in DNA replication and repair. GADD 45 stimulates DNA excision repair in vitro and inhibits entry ofcellsinto S phase. Thus, it has been suggested that GADD 45 mayserve as a link between the p53-dependentcellcycle checkpoint and DNA repair. GADD 45-like proteins, GADD 45 beta and GADD 45 gamma, have been shown to be induced by environmentalstresses. GADD 45 beta and GADD 45 gamma are thought to induce p38/JNK activation via MEKK4 activation.
Specifications
| GADD45B | |
| Polyclonal | |
| Unconjugated | |
| GADD45B | |
| AI323528; DKFZP566B133; Gadd45b; GADD45BETA; growth arrest and DNA damage inducible beta; growth arrest and DNA damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible 45 beta; growth arrest and DNA-damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible, beta; MYD118; myeloid differentiation primary response; myeloid differentiation primary response gene 118; myeloid differentiation primary response protein MyD118; Negative growth regulatory protein MyD118; Unknown (protein for MGC:138134) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17873, 4616 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75293, P22339 | |
| GADD45B | |
| Synthetic peptide directed towards the middle region of human GADD45B. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction