Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986896
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6896 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6896 Supplier Bioss Supplier No. BS5904R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cellcycle progression issubject to arrest at G1 and G2 checkpointsin response to DNA damage, presumablyto allow time for DNA repair prior to entryinto S andMphase, respectively. The p53 tumorsuppressor isrequired for one such G1 checkpoint and functionsto upregulate expression of GADD 45 and p21. GADD 45 binds both Cdks and PCNA, a protein involved in DNA replication and repair. GADD 45 stimulates DNA excision repair in vitro and inhibits entry ofcellsinto S phase. Thus, it has been suggested that GADD 45 mayserve as a link between the p53-dependentcellcycle checkpoint and DNA repair. GADD 45-like proteins, GADD 45 beta and GADD 45 gamma, have been shown to be induced by environmentalstresses. GADD 45 beta and GADD 45 gamma are thought to induce p38/JNK activation via MEKK4 activation.

Specifications

Antigen GADD45B
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GADD45B
Gene Accession No. O75293
Gene Alias AI323528; BOS_7788; DKFZP566B133; Gadd45b; GADD45BETA; growth arrest and DNA damage inducible beta; Growth arrest and DNA damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible 45 beta; growth arrest and DNA-damage-inducible protein GADD45 beta; growth arrest and DNA-damage-inducible, beta; Myd118; myeloid differentiation primary response; myeloid differentiation primary response gene 118; Myeloid differentiation primary response protein MyD118; Negative growth regulatory protein MyD118
Gene Symbols GADD45B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GADD45B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4616
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.