Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD45G Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300285MFV610
Description
GADD45G Polyclonal antibody specifically detects GADD45G in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
GADD45G | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
CR6DNA damage-inducible transcript 2 protein, Cytokine-responsive protein CR6, DDIT-2, DDIT2GADD45-gamma, growth arrest and DNA damage-inducible protein GADD45 gamma, growth arrest and DNA-damage-inducible, gamma, GRP1717 kD | |
Produced in rabbits immunized with purified, recombinant Human GADD45G (Accession#: O95257; Met1-Glu159) | |
0.1 mL | |
Apoptosis, Cancer | |
10912 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction