Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD45G Antibody (OTI2F12), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271646AF750
Description
GADD45G Monoclonal specifically detects GADD45G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GADD45G | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GADD45G | |
| Full length recombinant protein of human GADD45G (NP_006696) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2F12 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CR6DNA damage-inducible transcript 2 protein, Cytokine-responsive protein CR6, DDIT-2, DDIT2GADD45-gamma, growth arrest and DNA damage-inducible protein GADD45 gamma, growth arrest and DNA-damage-inducible, gamma, GRP1717 kD | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer | |
| 10912.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction