Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD45G Monoclonal Antibody (OTI3A1), TrueMAB™, OriGene
SDP

Catalog No. 502539716
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9716 30 μL
50-167-5899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9716 Supplier OriGene Supplier No. TA505421S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GADD45G Monoclonal antibody specifically detects GADD45G in Human samples. It is validated for Western Blot

Specifications

Antigen GADD45G
Applications Western Blot
Classification Monoclonal
Clone OTI3A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DCPS
Gene Accession No. Q96C86
Gene Alias AI327420; C86281; CR6; Cytokine-responsive protein CR6; DDIT2; DDIT-2; DNA damage-inducible transcript 2 protein; GADD45G; GADD45gamma; GADD45-gamma; gadd-related protein, 17 kD; growth arrest and DNA damage inducible; growth arrest and DNA damage inducible gamma; growth arrest and DNA damage-inducible protein GADD45 gamma; growth arrest and DNA-damage-inducible 45 gamma; growth arrest and DNA-damage-inducible protein GADD45 gamma; growth arrest and DNA-damage-inducible, gamma; GRP17; OIG37; RP11-260L6.1
Host Species Mouse
Immunogen Full length human recombinant protein of human DCPS produced in HEK293T cell
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28960
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.