Learn More
GADD45G Mouse anti-Human, Clone: OTI3A1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505421

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GADD45G gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45G is highly expressed in placenta.Specifications
GADD45G | |
Monoclonal | |
Unconjugated | |
GADD45G | |
CR6, DDIT2, GADD45gamma, GRP17 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10912 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3A1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95257 | |
GADD45G | |
Full length human recombit protein of human GADD45G produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.