Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GADD45G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100742
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100742 Supplier Invitrogen™ Supplier No. PA5100742
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GADD45G.

GADD45G gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The GADD45G is highly expressed in placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GADD45G
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GADD45G
Gene Accession No. O95257, Q9WTQ7, Q9Z111
Gene Alias AI327420; C86281; CR6; Cytokine-responsive protein CR6; DDIT2; DDIT-2; DNA damage-inducible transcript 2 protein; GADD45G; GADD45gamma; GADD45-gamma; gadd-related protein, 17 kD; growth arrest and DNA damage inducible; growth arrest and DNA damage inducible gamma; growth arrest and DNA damage-inducible protein GADD45 gamma; growth arrest and DNA-damage-inducible 45 gamma; growth arrest and DNA-damage-inducible protein GADD45 gamma; growth arrest and DNA-damage-inducible, gamma; GRP17; OIG37; RP11-260L6.1
Gene Symbols GADD45G
Host Species Rabbit
Immunogen A synthesized peptide derived from human GADD45G(Accession O95257), corresponding to amino acid residues A103-P153.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10912, 23882, 291005
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.