Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gAdiponectin/gAcrp30 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311025AF700
Description
gAdiponectin/gAcrp30 Polyclonal antibody specifically detects gAdiponectin/gAcrp30 in Human samples. It is validated for Western Blot, ELISASpecifications
gAdiponectin/gAcrp30 | |
Polyclonal | |
Western Blot, ELISA | |
Rabbit | |
Affinity purified | |
RUO | |
9370 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Recombinant Human gAdiponectin/gAcrp30 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction