Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Gadolinium

Catalog No. 503104459 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-310-4459 1 Ea.
1 options

Catalog No. 50-310-4459

Supplier: Sigma Aldrich GF283049511EA

Due to product restrictions, please to purchase or view availability for this product.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Gadolinium

Specifications

CAS 7440-54-2
Melting Point 1313°C
Density 7.886 g/mL (at 25°C)
Boiling Point 3273°C
Molecular Formula Gd
Linear Formula Gd
Quantity 1 Ea.
MDL Number MFCD00011022
Synonym GD006100
Molecular Weight (g/mol) 157.25
Percent Purity 99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.