Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Gadolinium chloride
SDP

Catalog No. 474150 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-415-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-415-0 Supplier R&D Systems™ Supplier No. 4741/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Stretch-activated calcium channel blocker; calcium-sensing receptor agonist

Calcium-sensing receptor (CaSR) agonist; induces NLRP3 inflammasome activation in bone marrow-derived macrophages. Blocks stretch-activated calcium channels; inhibits increase in intracellular calcium ion concentration in hypotonic-stimulated pulmonary artery smooth muscle cells. Has been demonstrated to block TRPML channel currents and other TRP channels.

Chemical Identifiers

CAS 10138-52-0
Molecular Formula Cl3Gd

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 263.61
Purity >98%
Molecular Weight (g/mol) 263.6
Chemical Name or Material Gadolinium chloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.