Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) nitrate hydrate, 99.5% (REO)

Catalog No. p-7049107
Click to view available options
Quantity:
25 g
100 g

CAS: 94219-55-3 | GdN3O9 | 343.26 g/mol

It is applied as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94219-55-3
Molecular Formula GdN3O9
Molecular Weight (g/mol) 343.26
MDL Number MFCD00011023
InChI Key MWFSXYMZCVAQCC-UHFFFAOYSA-N
IUPAC Name gadolinium(3+) trinitrate
SMILES [Gd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 92°C
Color White
Physical Form Crystalline
Quantity 25 g
UN Number UN1477
Merck Index 14,4326
Solubility Information Soluble in water.
Formula Weight 343.26 (Anhydrous)
Percent Purity 99.9%
Sensitivity Hygroscopic
Density 2.406 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
RTECSNumber LW4550000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.