Learn More
Gadolinium(III) nitrate hydrate, REacton™, 99.99% (REO)
Description
Gadolinium(III) nitrate is used as an oxidizing agent and the precursor material in the preparation of gadolinium compounds. It is extensively employed in the production of specialized glasses and ceramics. It is also employed in heavy water nuclear reactors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 92°C |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 25 g |
UN Number | UN1477 |
Merck Index | 14,4326 |
Solubility Information | Soluble in water. |
Formula Weight | 343.26 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H315-H319-H335 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
RTECSNumber | LW4550000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.