Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gadolinium(III) nitrate pentahydrate, 99.9%

Catalog No. p-222420
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 52788-53-1 | GdN3O9·5H2O | 433.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52788-53-1
Molecular Formula GdN3O9·5H2O
Molecular Weight (g/mol) 433.33
MDL Number MFCD00149729

Specifications

Melting Point 92.0°C
Color White
Physical Form Crystals
Quantity 25 g
Assay Percent Range 99.9%
Linear Formula Gd(NO3)3·5H2O
Merck Index 14, 4326
Formula Weight 433.33
Percent Purity 99.9%
Packaging Glass bottle
Chemical Name or Material Gadolinium(III) nitrate pentahydrate, 99.9%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.